(S)-(-)-1,1'-Bi(2-naphthol), 99%
(S)-(-)-1,1'-Bi(2-naphthol), 99%
(S)-(-)-1,1'-Bi(2-naphthol), 99%
Thermo Scientific Chemicals

(S)-(-)-1,1'-Bi(2-naphthol), 99%

CAS: 18531-99-2 | C20H14O2 | 286.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L08423.14
also known as L08423-14
Price (USD)
963.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,134.00
Save 170.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
963.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,134.00
Save 170.35 (15%)
Each
Chemical Identifiers
CAS18531-99-2
IUPAC Name[1,1'-binaphthalene]-2,2'-diol
Molecular FormulaC20H14O2
InChI KeyPPTXVXKCQZKFBN-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1C1=C(O)C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)205.0-212.0?C
Appearance (Color)White to pale cream to pale brown
Optical Rotation-35.0 ± 1.0? (C=1 in THF)
Assay (Silylated GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only