5-Norbornene-2-carboxylic acid, predominantly endo isomer, 97%
5-Norbornene-2-carboxylic acid, predominantly endo isomer, 97%
5-Norbornene-2-carboxylic acid, predominantly endo isomer, 97%
Thermo Scientific Chemicals

5-Norbornene-2-carboxylic acid, predominantly endo isomer, 97%

CAS: 120-74-1 | C8H10O2 | 138.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08444.06
also known as L08444-06
Price (USD)
28.65
Online Exclusive
31.80
Save 3.15 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
28.65
Online Exclusive
31.80
Save 3.15 (10%)
Each
Chemical Identifiers
CAS120-74-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%%
Refractive Index1.4915-1.4965 @ 20?C
Assay (GC)≥96.0%
FormLiquid or liquid as melt
5-Norbornene-2-carboxylic acid is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Norbornene-2-carboxylic acid is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible with water.

Notes
Incompatible with oxidizing agents, bases. Store away from oxidizing agents.
RUO – Research Use Only
Benjamin D Fairbanks; Michael P Schwartz; Alexandra E Halevi; Charles R Nuttelman; Christopher N Bowman; Kristi S Anseth. A Versatile Synthetic Extracellular Matrix Mimic via Thiol-Norbornene Photopolymerization.Advanced Materials. 2009, 21 (48), 5005-5010.