2-(2-Ethoxyethoxy)ethyl bromide, 98%, stab. with potassium carbonate
2-(2-Ethoxyethoxy)ethyl bromide, 98%, stab. with potassium carbonate
2-(2-Ethoxyethoxy)ethyl bromide, 98%, stab. with potassium carbonate
2-(2-Ethoxyethoxy)ethyl bromide, 98%, stab. with potassium carbonate
Thermo Scientific Chemicals

2-(2-Ethoxyethoxy)ethyl bromide, 98%, stab. with potassium carbonate

CAS: 54550-36-6 | C6H13BrO2 | 197.072 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L08522.18
also known as L08522-18
Price (USD)
747.65
Online Exclusive
830.00
Save 82.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
747.65
Online Exclusive
830.00
Save 82.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS54550-36-6
IUPAC Name1-bromo-2-(2-ethoxyethoxy)ethane
Molecular FormulaC6H13BrO2
InChI KeyUEDIWIFQWRXXJG-UHFFFAOYSA-N
SMILESCCOCCOCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid containing suspended powder
Refractive Index1.4510-1.4550 @ 20?C
Assay (GC)>97.5%
2-(2-Ethoxyethoxy)ethyl bromide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Ethoxyethoxy)ethyl bromide is used as pharmaceutical intermediates.

Solubility
Not miscible with water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only