Sarcosine methyl ester hydrochloride, 97%
Sarcosine methyl ester hydrochloride, 97%
Sarcosine methyl ester hydrochloride, 97%
Sarcosine methyl ester hydrochloride, 97%
Thermo Scientific Chemicals

Sarcosine methyl ester hydrochloride, 97%

CAS: 13515-93-0 | C4H10ClNO2 | 139.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L08572.09
also known as L08572-09
Price (USD)
46.65
Online Exclusive
51.90
Save 5.25 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
51.90
Save 5.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS13515-93-0
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤1.0% (70?C/constant weight)
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (Non-aqueous acid-base Titration)≥96.0%
It can react with formic acid to get N-formyl-N-methyl-glycine methyl ester in the presence of HCO2Na at ambient temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can react with formic acid to get N-formyl-N-methyl-glycine methyl ester in the presence of HCO2Na at ambient temperature.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Dejian Zhou, et al. Langmuir-Blodgett Films and Photophysical Properties of a C60-Sarcosine Methyl Ester Derivative, C60(C5H9NO2).J. Phys. Chem.,1996,100(8), 3150-3156.
  2. M. Sheinblatt. NMR Study of the Protolysis Kinetics in Simple Amino Acids. I. Sarcosine Hydrochloride and its Ester.J. Chem. Phys.,1962,363103.