(1-Hexyl)trimethylammonium bromide, 98%
(1-Hexyl)trimethylammonium bromide, 98%
(1-Hexyl)trimethylammonium bromide, 98%
Thermo Scientific Chemicals

(1-Hexyl)trimethylammonium bromide, 98%

CAS: 2650-53-5 | C9H22BrN | 224.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08594.06
also known as L08594-06
Price (USD)
39.65
Online Exclusive
43.70
Save 4.05 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.70
Save 4.05 (9%)
Each
Chemical Identifiers
CAS2650-53-5
IUPAC Namehexyltrimethylazanium bromide
Molecular FormulaC9H22BrN
InChI KeyJYVPKRHOTGQJSE-UHFFFAOYSA-M
SMILES[Br-].CCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water Content (Karl Fischer Titration)<2.5%
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Melting Point (clear melt)182-187?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only