Trimethyl-1-octylammonium bromide, 97%
Trimethyl-1-octylammonium bromide, 97%
Trimethyl-1-octylammonium bromide, 97%
Thermo Scientific Chemicals

Trimethyl-1-octylammonium bromide, 97%

CAS: 2083-68-3 | C11H26BrN | 252.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L08610.06
also known as L08610-06
Price (USD)
39.65
Online Exclusive
43.70
Save 4.05 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.70
Save 4.05 (9%)
Each
Chemical Identifiers
CAS2083-68-3
IUPAC Nametrimethyl(octyl)azanium bromide
Molecular FormulaC11H26BrN
InChI KeyXCOHAFVJQZPUKF-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Bromide)≥96.0 to ≤104.0%
Water Content (Karl Fischer Titration)≤2.25%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or lumps or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only