(S)-(+)-S-Methyl-S-phenylsulfoximine, 97%
(S)-(+)-S-Methyl-S-phenylsulfoximine, 97%
(S)-(+)-S-Methyl-S-phenylsulfoximine, 97%
(S)-(+)-S-Methyl-S-phenylsulfoximine, 97%
Thermo Scientific Chemicals

(S)-(+)-S-Methyl-S-phenylsulfoximine, 97%

CAS: 33903-50-3 | C7H9NOS | 155.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L08618.MD
also known as L08618-MD
Price (USD)
80.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.80
Save 14.15 (15%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.80
Save 14.15 (15%)
Each
Add to cart
Chemical Identifiers
CAS33903-50-3
IUPAC Nameimino(methyl)phenyl-λ⁶-sulfanone
Molecular FormulaC7H9NOS
InChI KeyYFYIDTVGWCYSEO-UHFFFAOYNA-N
SMILESCS(=N)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or fused solid or clear liquid or viscous liquid as melt
Optical Rotation34.9° to 36.9° (c=1 in acetone)
Assay (GC)≥96.0%
Melting Point (clear melt)25.0-34.0°C
Appearance (Color)Colorless to white to pale cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only