2,2,2-Trifluoroethyl trichloromethanesulfonate, 94%
2,2,2-Trifluoroethyl trichloromethanesulfonate, 94%
2,2,2-Trifluoroethyl trichloromethanesulfonate, 94%
Thermo Scientific Chemicals

2,2,2-Trifluoroethyl trichloromethanesulfonate, 94%

CAS: 23199-56-6 | C3H2Cl3F3O3S | 281.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L08635.14
also known as L08635-14
Price (USD)
475.65
Special offer
Online exclusive
Ends: 30-Jun-2026
559.00
Save 83.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
475.65
Special offer
Online exclusive
Ends: 30-Jun-2026
559.00
Save 83.35 (15%)
Each
Chemical Identifiers
CAS23199-56-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4185-1.4255 @ 20°C
FormLiquid
Assay (GC)≥92.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only