Dimethyl 5-norbornene-2,3-dicarboxylate, 94%
Dimethyl 5-norbornene-2,3-dicarboxylate, 94%
Dimethyl 5-norbornene-2,3-dicarboxylate, 94%
Thermo Scientific Chemicals

Dimethyl 5-norbornene-2,3-dicarboxylate, 94%

CAS: 5826-73-3 | C11H14O4 | 210.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L08664.18
also known as L08664-18
Price (USD)
328.65
Online Exclusive
365.00
Save 36.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
328.65
Online Exclusive
365.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS5826-73-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥92.0%
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4830-1.4900 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only