(S)-(+)-2-Hydroxy-2-phenylpropionic acid, 98+%
(S)-(+)-2-Hydroxy-2-phenylpropionic acid, 98+%
(S)-(+)-2-Hydroxy-2-phenylpropionic acid, 98+%
Thermo Scientific Chemicals

(S)-(+)-2-Hydroxy-2-phenylpropionic acid, 98+%

CAS: 13113-71-8 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L08692.MD
also known as L08692-MD
Price (USD)
75.65
Online Exclusive
83.80
Save 8.15 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
83.80
Save 8.15 (10%)
Each
Chemical Identifiers
CAS13113-71-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)112.0-119.0?C
Optical Rotation49.5 ± 0.4? (C=2 in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only