(R)-(-)-alpha-Methoxyphenylacetic acid, 99%
(R)-(-)-alpha-Methoxyphenylacetic acid, 99%
(R)-(-)-alpha-Methoxyphenylacetic acid, 99%
Thermo Scientific Chemicals

(R)-(-)-alpha-Methoxyphenylacetic acid, 99%

CAS: 3966-32-3 | C9H10O3 | 166.176 g/mol
Have Questions?
Catalog NumberQuantity
L08719.MD
also known as L08719-MD
250 mg
Catalog number L08719.MD
also known as L08719-MD
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.20
Save 6.55 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.20
Save 6.55 (14%)
Each
Chemical Identifiers
CAS3966-32-3
IUPAC Name(2R)-2-methoxy-2-phenylacetate
Molecular FormulaC9H9O3
InChI KeyDIWVBIXQCNRCFE-MRVPVSSYSA-M
SMILESCO[C@@H](C([O-])=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay (Silylated GC)≥98.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only