N,N-Dimethylsuccinamic acid, 98%
N,N-Dimethylsuccinamic acid, 98%
N,N-Dimethylsuccinamic acid, 98%
Thermo Scientific Chemicals

N,N-Dimethylsuccinamic acid, 98%

CAS: 2564-95-6 | C6H11NO3 | 145.158 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08796.06
also known as L08796-06
Price (USD)
69.65
Online exclusive
76.70
Save 7.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
69.65
Online exclusive
76.70
Save 7.05
Each
Specifications
Beilstein1762788
CAS2564-95-6
Chemical Name or MaterialN,N-Dimethylsuccinamic acid
EINECS Number000-000-0
Formula Weight145.16
View more
It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Galvin M. Coppinger. Preparations of N,N-Dimethylamides. J. Am. Chem. Soc. 1954, 76 (5), 1372-1373.
  2. David Pressman; John H. Bryden; Linus Pauling. The Reactions of Antiserum Homologous to the p-Azosuccinanilate Ion Group1a. J. Am. Chem. Soc. 1948, 70(4), 1352-1358.