It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Galvin M. Coppinger. Preparations of N,N-Dimethylamides. J. Am. Chem. Soc. 1954, 76 (5), 1372-1373.
- David Pressman; John H. Bryden; Linus Pauling. The Reactions of Antiserum Homologous to the p-Azosuccinanilate Ion Group1a. J. Am. Chem. Soc. 1948, 70(4), 1352-1358.