n-Butylcyclohexane, 99%
n-Butylcyclohexane, 99%
n-Butylcyclohexane, 99%
Thermo Scientific Chemicals

n-Butylcyclohexane, 99%

CAS: 1678-93-9 | C10H20 | 140.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L08845.09
also known as L08845-09
Price (USD)
35.10
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
35.10
Each
Chemical Identifiers
CAS1678-93-9
IUPAC Namebutylcyclohexane
Molecular FormulaC10H20
InChI KeyGGBJHURWWWLEQH-UHFFFAOYSA-N
SMILESCCCCC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.4395-1.4425 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only