2,3-Dichloro-1-propene, 98%
2,3-Dichloro-1-propene, 98%
2,3-Dichloro-1-propene, 98%
Thermo Scientific Chemicals

2,3-Dichloro-1-propene, 98%

CAS: 78-88-6 | C3H4Cl2 | 110.965 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L08943.18
also known as L08943-18
Price (USD)
74.65
Online Exclusive
83.10
Save 8.45 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
83.10
Save 8.45 (10%)
Each
Chemical Identifiers
CAS78-88-6
IUPAC Name2,3-dichloroprop-1-ene
Molecular FormulaC3H4Cl2
InChI KeyFALCMQXTWHPRIH-UHFFFAOYSA-N
SMILESClCC(Cl)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow to pale brown
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4595-1.4635 @ 20?C
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (2g/L).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. T Neudecker and D Henschler. Mutagenicity of chloroolefins in the Salmonella/mammalian microsome test. III. Metabolic activation of the allylic chloropropenes allyl chloride, 1,3-dichloropropene, 2,3-dichloro-1-propene, 1,2,3-trichloropropene, 1,1,2,3-tetrachloro-2-propene and hexachloropropene by S9 mix via two different metabolic pathways. Mutation Research/Fundamental and Molecular Mechanisms of Mutagenesis,1986, 107 (1-2), 1-9.
  2. Roland P H Schmitz et. al. Evidence for a radical mechanism of the dechlorination of chlorinated propenes mediated by the tetrachloroethene reductive dehalogenase of Sulfurospirillum muftivorans. Environmental Science & Technology. 2007, 41 (21), 7370-7375.
  3. Reductive elimination with Zn powder gives allene free from methylacetylene: Org. Synth. Coll., 5, 22 (1973).
  4. Electron-acceptor, with concomitant reduction to allene, in a high-yield coupling of aryl Grignard reagents to symmetrical biaryls: Tetrahedron Lett., 29, 1293 (1988):