n-Propyl 4-aminobenzoate, 98%
n-Propyl 4-aminobenzoate, 98%
n-Propyl 4-aminobenzoate, 98%
Thermo Scientific Chemicals

n-Propyl 4-aminobenzoate, 98%

CAS: 94-12-2 | C10H13NO2 | 179.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L09066.18
also known as L09066-18
Price (USD)
108.65
Special offer
Online exclusive
Ends: 30-Jun-2026
128.00
Save 19.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
108.65
Special offer
Online exclusive
Ends: 30-Jun-2026
128.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS94-12-2
IUPAC Namepropyl 4-aminobenzoate
Molecular FormulaC10H13NO2
InChI KeyNBFQYHKHPBMJJV-UHFFFAOYSA-N
SMILESCCCOC(=O)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)71.0-77.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only