2-Chlorobenzenesulfonamide, 98%
2-Chlorobenzenesulfonamide, 98%
2-Chlorobenzenesulfonamide, 98%
Thermo Scientific Chemicals

2-Chlorobenzenesulfonamide, 98%

CAS: 6961-82-6 | C6H6ClNO2S | 191.629 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09082.03
also known as L09082-03
Price (USD)
23.65
Special offer
Online exclusive
Ends: 30-Jun-2026
27.20
Save 3.55 (13%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
23.65
Special offer
Online exclusive
Ends: 30-Jun-2026
27.20
Save 3.55 (13%)
Each
Chemical Identifiers
CAS6961-82-6
IUPAC Name2-chlorobenzene-1-sulfonamide
Molecular FormulaC6H6ClNO2S
InChI KeyJCCBZCMSYUSCFM-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Melting Point (clear melt)187.0-193.0?C
Appearance (Color)White to dark yellow to pale yellow
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only