Diethyl tert-butylmalonate, 96%
Diethyl tert-butylmalonate, 96%
Diethyl tert-butylmalonate, 96%
Thermo Scientific Chemicals

Diethyl tert-butylmalonate, 96%

CAS: 759-24-0 | C11H20O4 | 216.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09131.14
also known as L09131-14
Price (USD)
490.65
Online Exclusive
545.00
Save 54.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
490.65
Online Exclusive
545.00
Save 54.35 (10%)
Each
Chemical Identifiers
CAS759-24-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4225-1.4275 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
FormLiquid
Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Patel, N. C.; Schwarz, J. B.; Islam, K.; Miller, W.; Tran, T. P.; Wei, Y. Improved Synthesis of gamma-Lactones from Cyclopropyl Cyanoesters. Synth. Commun. 2011, 41 (15), 2209-2215.
  2. Huang, J. Y.; Zhuang, J. M.; Zou, Y. S. Substituent effects of accelerator on nitroxide-mediated radical polymerization. Macromol. Res. 2011, 19 (8), 757-763.