3-Fluorobenzoyl chloride, 98%
3-Fluorobenzoyl chloride, 98%
3-Fluorobenzoyl chloride, 98%
Thermo Scientific Chemicals

3-Fluorobenzoyl chloride, 98%

CAS: 1711-07-5 | C7H4ClFO | 158.556 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09152.14
also known as L09152-14
Price (USD)
80.65
Online Exclusive
89.90
Save 9.25 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.90
Save 9.25 (10%)
Each
Chemical Identifiers
CAS1711-07-5
IUPAC Name3-fluorobenzoyl chloride
Molecular FormulaC7H4ClFO
InChI KeySYVNVEGIRVXRQH-UHFFFAOYSA-N
SMILESFC1=CC=CC(=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5265-1.5305 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only