6-Methyl-3,5-heptadien-2-one, 97%
6-Methyl-3,5-heptadien-2-one, 97%
6-Methyl-3,5-heptadien-2-one, 97%
Thermo Scientific Chemicals

6-Methyl-3,5-heptadien-2-one, 97%

CAS: 1604-28-0 | C8H12O | 124.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09169.06
also known as L09169-06
Price (USD)
37.65
Online Exclusive
41.30
Save 3.65 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.30
Save 3.65 (9%)
Each
Chemical Identifiers
CAS1604-28-0
IUPAC Name(3E)-6-methylhepta-3,5-dien-2-one
Molecular FormulaC8H12O
InChI KeyKSKXSFZGARKWOW-GQCTYLIASA-N
SMILESCC(C)=CC=CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5315-1.5365 @ 20?C
6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-3,5-heptadien-2-one is used in the preparation of (RS)-(E)-2,6-dimethyl-1,2-epoxy-3,5-heptadiene.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Joshi, R.; Gulati, A. Fractionation and identification of minor and aroma-active constituents in Kangra orthodox black tea. Food Chem. 2015, 167, 290-298.
  2. Schepetkin, I. A.; Kushnarenko, S. V.; Özek, G.; Kirpotina, L. N.; Utegenova, G. A.; Kotukhov, Y. A.; Danilova, A. N.; Özek, T.; Başer, K. H. C.; Quinn, M. T. Inhibition of Human Neutrophil Responses by the Essential Oil of Artemisia kotuchovii and Its Constituents. J. Agric. Food Chem. 2015, 63 (20), 4999-5007.