3-Phenylpropiophenone, 98%
3-Phenylpropiophenone, 98%
3-Phenylpropiophenone, 98%
Thermo Scientific Chemicals

3-Phenylpropiophenone, 98%

CAS: 1083-30-3 | C15H14O | 210.276 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L09173.09
also known as L09173-09
Price (USD)
102.65
Special offer
Online exclusive
Ends: 30-Jun-2026
121.00
Save 18.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 30-Jun-2026
121.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS1083-30-3
IUPAC Name1,3-diphenylpropan-1-one
Molecular FormulaC15H14O
InChI KeyQGGZBXOADPVUPN-UHFFFAOYSA-N
SMILESO=C(CCC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormCrystalline powder
Assay (GC)>97.5%
Clarity(5% w/v solution in acetone) : Transparent to almost transparent by JIS K 8001 5.2
3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
Heating with excess SOCl2 and a catalytic amount of pyridine gives 2-benzoyl-3-chlorobenzo[b]thiophene: J. Org. Chem., 41, 3399 (1976). For similar reactions, see trans-Cinnamic acid, A13538.