N-Boc-D-valine, 98+%
N-Boc-D-valine, 98+%
N-Boc-D-valine, 98+%
Thermo Scientific Chemicals

N-Boc-D-valine, 98+%

CAS: 22838-58-0 | C10H19NO4 | 217.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09193.03
also known as L09193-03
Price (USD)
27.65
Online Exclusive
30.50
Save 2.85 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.50
Save 2.85 (9%)
Each
Chemical Identifiers
CAS22838-58-0
IUPAC Name(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
Molecular FormulaC10H19NO4
InChI KeySZXBQTSZISFIAO-SSDOTTSWSA-N
SMILESCC(C)[C@@H](NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation6.5 ± 0.4? (c=1 in acetic acid)
Melting Point (clear melt)75.0-81.0?C
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only