4-(Methylthio)phenyl isothiocyanate, 97%
4-(Methylthio)phenyl isothiocyanate, 97%
4-(Methylthio)phenyl isothiocyanate, 97%
4-(Methylthio)phenyl isothiocyanate, 97%
Thermo Scientific Chemicals

4-(Methylthio)phenyl isothiocyanate, 97%

CAS: 15863-41-9 | C8H7NS2 | 181.271 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09270.06
also known as L09270-06
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS15863-41-9
IUPAC Name1-isothiocyanato-4-(methylsulfanyl)benzene
Molecular FormulaC8H7NS2
InChI KeyCEBAJHCAFXYWNT-UHFFFAOYSA-N
SMILESCSC1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)51.5-60.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only