1,1-Diphenylpropane, 98+%
1,1-Diphenylpropane, 98+%
1,1-Diphenylpropane, 98+%
Thermo Scientific Chemicals

1,1-Diphenylpropane, 98+%

CAS: 1530-03-6 | C15H16 | 196.29 g/mol
Have Questions?
Catalog NumberQuantity
L09274.06
also known as L09274-06
5 g
Catalog number L09274.06
also known as L09274-06
Price (USD)
69.65
Online Exclusive
77.40
Save 7.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
77.40
Save 7.75 (10%)
Each
Chemical Identifiers
CAS1530-03-6
IUPAC Name(1-phenylpropyl)benzene
Molecular FormulaC15H16
InChI KeyBUZMJVBOGDBMGI-UHFFFAOYSA-N
SMILESCCC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
GC/MS AnalysisConforms
Assay (GC)≥98.5%
Refractive Index1.564
Appearance (Color)Colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only