(S)-(+)-2-Pentanol, 97%
(S)-(+)-2-Pentanol, 97%
(S)-(+)-2-Pentanol, 97%
Thermo Scientific Chemicals

(S)-(+)-2-Pentanol, 97%

CAS: 26184-62-3 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L09314.MD
also known as L09314-MD
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.90
Save 12.25 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.90
Save 12.25 (14%)
Each
Specifications
Beilstein1718820
CAS26184-62-3
Chemical Name or Material(S)-(+)-2-Pentanol
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
Density0.81
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only