2-Fluorobenzophenone, 98+%
2-Fluorobenzophenone, 98+%
2-Fluorobenzophenone, 98+%
Thermo Scientific Chemicals

2-Fluorobenzophenone, 98+%

CAS: 342-24-5 | C13H9FO | 200.212 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09316.14
also known as L09316-14
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
121.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS342-24-5
IUPAC Name(2-fluorophenyl)(phenyl)methanone
Molecular FormulaC13H9FO
InChI KeyDWFDQVMFSLLMPE-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5850-1.5890 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only