2-Pentadecanone, 97%
2-Pentadecanone, 97%
2-Pentadecanone, 97%
Thermo Scientific Chemicals

2-Pentadecanone, 97%

CAS: 2345-28-0 | C15H30O | 226.404 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L09347.22
also known as L09347-22
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS2345-28-0
IUPAC Namepentadecan-2-one
Molecular FormulaC15H30O
InChI KeyCJPNOLIZCWDHJK-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥ 96.0 %
Melting Point37- 41?C
Infrared spectrumConforms
Solution TestColor: Colorless
Solution TestTurbidity (5% w/w in 95% Ethanol): Clear
View more
It is primarily used as a flavoring substances to impart a fatty, slightly spicy, and floral flavor. 2-Pentadecanone can perfectly compliment and enhance a variety of foods including butters, cheeses and meats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is primarily used as a flavoring substances to impart a fatty, slightly spicy, and floral flavor. 2-Pentadecanone can perfectly compliment and enhance a variety of foods including butters, cheeses and meats.

Solubility
Soluble in toluene (almost transparency). Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Eriko Harada; Jun Nakagawa; Tsunaki Asano; Masato Taoka; Hiroyuki Sorimachi; Yoshihiro Ito; Toshiro Aigaki; Takashi Matsuo.Functional evolution of duplicated odorant-binding protein genes, Obp57d and Obp57e, in Drosophila. PLoS ONE,2012, 1 (7), e29710.
  2. Angela M. Schaner; Liane D. Tanico-Hogan; Larry L. Jackson. (S)-2-pentadecyl acetate and 2-pentadecanone Components of aggregation pheromone ofDrosophila busckii. Journal of Chemical Ecology,1989, 15(11),2577-2588.