4-Hydroxyphenyl benzoate, 98%
4-Hydroxyphenyl benzoate, 98%
4-Hydroxyphenyl benzoate, 98%
4-Hydroxyphenyl benzoate, 98%
Thermo Scientific Chemicals

4-Hydroxyphenyl benzoate, 98%

CAS: 2444-19-1 | C13H10O3 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09350.14
also known as L09350-14
Price (USD)
232.65
Special offer
Online exclusive
Ends: 30-Jun-2026
274.00
Save 41.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
232.65
Special offer
Online exclusive
Ends: 30-Jun-2026
274.00
Save 41.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS2444-19-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Melting Point (clear melt)162.0-168.0?C
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only