4-Ethylcyclohexanone, 99%
4-Ethylcyclohexanone, 99%
4-Ethylcyclohexanone, 99%
Thermo Scientific Chemicals

4-Ethylcyclohexanone, 99%

CAS: 5441-51-0 | C8H14O | 126.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L09363.14
also known as L09363-14
Price (USD)
61.65
Online exclusive
68.30
Save 6.65
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
61.65
Online exclusive
68.30
Save 6.65
Each
Chemical Identifiers
CAS5441-51-0
IUPAC Name4-ethylcyclohexan-1-one
Molecular FormulaC8H14O
InChI KeyOKSDJGWHKXFVME-UHFFFAOYSA-N
SMILESCCC1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Refractive Index1.4505-1.4535 @ 20°C
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only