N-Benzylthiourea, 98%
N-Benzylthiourea, 98%
N-Benzylthiourea, 98%
Thermo Scientific Chemicals

N-Benzylthiourea, 98%

CAS: 621-83-0 | C8H10N2S | 166.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L09400.06
also known as L09400-06
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.80
Save 6.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.80
Save 6.15
Each
Chemical Identifiers
CAS621-83-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Assay (HPLC)≥97.5%
Melting Point (clear melt)160.0-166.0?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only