(S)-(-)-2-Chloropropionic acid, 98%
(S)-(-)-2-Chloropropionic acid, 98%
(S)-(-)-2-Chloropropionic acid, 98%
Thermo Scientific Chemicals

(S)-(-)-2-Chloropropionic acid, 98%

CAS: 29617-66-1 | C3H4ClO2 | 107.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09427.06
also known as L09427-06
Price (USD)
62.65
Online Exclusive
68.90
Save 6.25 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
62.65
Online Exclusive
68.90
Save 6.25 (9%)
Each
Chemical Identifiers
CAS29617-66-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow to yellow to brown
FormLiquid or viscous liquid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Identification (FTIR)Conforms
View more
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

Solubility
Fully miscible in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ioannis N. Houpis.; Lawrence E. Patterson.; Charles A. Alt.; John R. Rizzo.; Tony Y. Zhang.; Michael Haurez. Synthesis of PPAR Agonist via Asymmetric Hydrogenation of a Cinnamic Acid Derivative and Stereospecific Displacement of (S)-2-Chloropropionic Acid.Org. Lett. 2005, 7 (10),1947-1950 .
  2. Atsushi Kurata.; Tatsuo Kurihara.; Harumi Kamachi.; Nobuyoshi Esaki. Asymmetric reduction of 2-chloroacrylic acid to (S)-2-chloropropionic acid by a novel reductase from Burkholderia sp. WSTetrahedron: Asymmetry. 2004, 15 (18),2837-2839 .