(R)-(-)-2-Phenylbutyric acid, 99%
(R)-(-)-2-Phenylbutyric acid, 99%
(R)-(-)-2-Phenylbutyric acid, 99%
Thermo Scientific Chemicals

(R)-(-)-2-Phenylbutyric acid, 99%

CAS: 938-79-4 | C10H12O2 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09464.06
also known as L09464-06
Price (USD)
323.65
Online Exclusive
359.00
Save 35.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
323.65
Online Exclusive
359.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS938-79-4
IUPAC Name2-phenylbutanoic acid
Molecular FormulaC10H12O2
InChI KeyOFJWFSNDPCAWDK-UHFFFAOYNA-N
SMILESCCC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)>98.5%
Refractive Index1.5135 - 1.5165
Appearance (Color)Clear, colourless
Assay (Silylated GC)>98.5%
Formliquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only