Ethyl 3,3-diethoxypropionate, 96%
Ethyl 3,3-diethoxypropionate, 96%
Ethyl 3,3-diethoxypropionate, 96%
Thermo Scientific Chemicals

Ethyl 3,3-diethoxypropionate, 96%

CAS: 10601-80-6 | C9H18O4 | 190.239 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L09562.09
also known as L09562-09
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.90
Save 8.25 (14%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.90
Save 8.25 (14%)
Each
Chemical Identifiers
CAS10601-80-6
IUPAC Nameethyl 3,3-diethoxypropanoate
Molecular FormulaC9H18O4
InChI KeySIALOQYKFQEKOG-UHFFFAOYSA-N
SMILESCCOC(CC(=O)OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 95.0%
Appearance (Color)Clear, colourless
Refractive Index1.4090 - 1.4150 @20?C
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only