(S)-(+)-2-(Methoxymethyl)pyrrolidine, 98%
(S)-(+)-2-(Methoxymethyl)pyrrolidine, 98%
(S)-(+)-2-(Methoxymethyl)pyrrolidine, 98%
Thermo Scientific Chemicals

(S)-(+)-2-(Methoxymethyl)pyrrolidine, 98%

CAS: 63126-47-6 | C6H13NO | 115.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09608.06
also known as L09608-06
Price (USD)
247.65
Online Exclusive
275.00
Save 27.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
247.65
Online Exclusive
275.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS63126-47-6
IUPAC Name2-(methoxymethyl)pyrrolidine
Molecular FormulaC6H13NO
InChI KeyCHPRFKYDQRKRRK-UHFFFAOYNA-N
SMILESCOCC1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Optical Rotation37.4 ± 0.1? (C=2 in 1N HCl)
Appearance (Color)Clear colorless to yellow
Refractive Index1.4440-1.4490 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only