N-(n-Butyl)thiourea, 98%
N-(n-Butyl)thiourea, 98%
N-(n-Butyl)thiourea, 98%
Thermo Scientific Chemicals

N-(n-Butyl)thiourea, 98%

CAS: 1516-32-1 | C5H12N2S | 132.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L09654.09
also known as L09654-09
Price (USD)
207.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
207.00
Each
Chemical Identifiers
CAS1516-32-1
IUPAC Namebutylthiourea
Molecular FormulaC5H12N2S
InChI KeyGMEGXJPUFRVCPX-UHFFFAOYSA-N
SMILESCCCCNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)73.5-79.5?C
FormCrystals or powder or crystalline powder or granules
Appearance (Color)White to pale cream
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only