1-Hexanethiol, 96%
1-Hexanethiol, 96%
1-Hexanethiol, 96%
Thermo Scientific Chemicals

1-Hexanethiol, 96%

CAS: 111-31-9 | C6H14S | 118.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number L09659.AC
also known as L09659-AC
Price (USD)
42.50
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
42.50
Each
Chemical Identifiers
CAS111-31-9
IUPAC Namehexane-1-thiol
Molecular FormulaC6H14S
InChI KeyPMBXCGGQNSVESQ-UHFFFAOYSA-N
SMILESCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥95.0%
Refractive Index1.4460-1.4520 @ 20°C (non-U.S. specification)
FormLiquid
Identification (FTIR)Conforms (non-U.S. specification)
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

Solubility
Difficult to mix in water.

Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Edith Chow.; Thomas R. Gengenbach.; Lech Wieczorek.; Burkhard Raguse. Detection of organics in aqueous solution using gold nanoparticles modified with mixed monolayers of 1-hexanethiol and 4-mercaptophenol.Sens. Actuators, B. 2010, 143 (2), 704-711 .
  2. Gary K. Rowe.; Stephen E. Creager. Chain Length and Solvent Effects on Competitive Self-Assembly of Ferrocenylhexanethiol and 1-Alkanethiols onto Gold. Langmuir. 1994, 10 (4),1186-1192 .