4-Aminobenzo-2,1,3-thiadiazole, 98%
4-Aminobenzo-2,1,3-thiadiazole, 98%
4-Aminobenzo-2,1,3-thiadiazole, 98%
Thermo Scientific Chemicals

4-Aminobenzo-2,1,3-thiadiazole, 98%

CAS: 767-64-6 | C6H5N3S | 151.187 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09678.03
also known as L09678-03
Price (USD)
65.90
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
65.90
Each
Chemical Identifiers
CAS767-64-6
IUPAC Name2,1,3-benzothiadiazol-4-amine
Molecular FormulaC6H5N3S
InChI KeyDRLGIZIAMHIQHL-UHFFFAOYSA-N
SMILESNC1=CC=CC2=NSN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow or Orange or Orange-yellow to brown
FormPowder or crystalline powder or crystals
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Melting Point (clear melt)66-70°C
4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Elizabeth C Banks; Stephen W Doughty; Steven M Toms; Richard T Wheelhouse; Anna Nicolaou. Inhibition of cobalamin-dependent methionine synthase by substituted benzo-fused heterocycles.FEBS Journal.2007, 274 (1), 287-299.
  2. N N Piatovskaia; I E Brumberg. [Comparative evaluation of the effectiveness of radioprotective agents judged from their protection of bone marrow and the gastrointestinal tract].Radiobiologiia.1982, 22 (2), 183-186.