2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%
Thermo Scientific Chemicals

2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%

CAS: 4274-38-8 | C7H7ClF3NS | 229.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09683.06
also known as L09683-06
Price (USD)
84.65
Online Exclusive
93.90
Save 9.25 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
84.65
Online Exclusive
93.90
Save 9.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS4274-38-8
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)188-204°C
Appearance (Color)White to cream to yellow or grey
FormPowder or crystalline powder or lumps
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only