3,5-Dimethylbenzaldehyde, 98%
3,5-Dimethylbenzaldehyde, 98%
3,5-Dimethylbenzaldehyde, 98%
3,5-Dimethylbenzaldehyde, 98%
Thermo Scientific Chemicals

3,5-Dimethylbenzaldehyde, 98%

CAS: 5779-95-3 | C9H10O | 134.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09748.03
also known as L09748-03
Price (USD)
49.65
Online Exclusive
54.80
Save 5.15 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.80
Save 5.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS5779-95-3
IUPAC Name3,5-dimethylbenzaldehyde
Molecular FormulaC9H10O
InChI KeyNBEFMISJJNGCIZ-UHFFFAOYSA-N
SMILESCC1=CC(C=O)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.5365-1.5405 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Free acid (titration)≤1.5%
3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzaldehyde is used to produce 2,3-bis-(3,5-dimethyl-phenyl)-succinonitrile by heating along with reagent NaCN and solvent H2O, methanol.

Solubility
Not miscible with water.

Notes
Handle and store under inert gas. Air Sensitive. Store away from air and oxidizing agents. Incompatible with oxidizing agents and air.
RUO – Research Use Only