It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Christopher J. Mortko.; Hung Dang.; Luis M. Campos.; Miguel A. Garcı́a-Garibay. H-abstraction prevails over α-cleavage in the solution and solid state photochemistry of cis-2,6-di(1-cyclohexenyl)cyclohexanone.Tetrahedron Lett. 2003, 44 (32),6133-6136 .