(+/-)-3-Phenyllactic acid, 98+%
(+/-)-3-Phenyllactic acid, 98+%
(+/-)-3-Phenyllactic acid, 98+%
Thermo Scientific Chemicals

(+/-)-3-Phenyllactic acid, 98+%

CAS: 828-01-3 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09854.06
also known as L09854-06
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS828-01-3
IUPAC Name2-hydroxy-3-phenylpropanoic acid
Molecular FormulaC9H10O3
InChI KeyVOXXWSYKYCBWHO-UHFFFAOYNA-N
SMILESOC(CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only