3-Bromophenyl isothiocyanate, 97%
3-Bromophenyl isothiocyanate, 97%
3-Bromophenyl isothiocyanate, 97%
Thermo Scientific Chemicals

3-Bromophenyl isothiocyanate, 97%

CAS: 2131-59-1 | C7H4BrNS | 214.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09871.14
also known as L09871-14
Price (USD)
161.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
161.00
Each
Chemical Identifiers
CAS2131-59-1
IUPAC Name1-bromo-3-isothiocyanatobenzene
Molecular FormulaC7H4BrNS
InChI KeyZMGMGHNOACSMQN-UHFFFAOYSA-N
SMILESBrC1=CC=CC(=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6810-1.6860 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only