5-Cyano-2-methylpyridine, 99%
5-Cyano-2-methylpyridine, 99%
5-Cyano-2-methylpyridine, 99%
5-Cyano-2-methylpyridine, 99%
Thermo Scientific Chemicals

5-Cyano-2-methylpyridine, 99%

CAS: 3222-48-8 | C7H6N2 | 118.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09875.03
also known as L09875-03
Price (USD)
33.50
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
33.50
Each
Add to cart
Chemical Identifiers
CAS3222-48-8
IUPAC Name6-methylpyridine-3-carbonitrile
Molecular FormulaC7H6N2
InChI KeyPBLOYQAQGYUPCM-UHFFFAOYSA-N
SMILESCC1=CC=C(C=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)White to pale cream to cream to yellow to brown
FormCrystals or powder or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only