Bis(2-dimethylaminoethyl) ether, 98%
Bis(2-dimethylaminoethyl) ether, 98%
Bis(2-dimethylaminoethyl) ether, 98%
Thermo Scientific Chemicals

Bis(2-dimethylaminoethyl) ether, 98%

CAS: 3033-62-3 | C8H20N2O | 160.26 g/mol
Have Questions?
Catalog NumberQuantity
L09884.18
also known as L09884-18
50 g
Catalog number L09884.18
also known as L09884-18
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS3033-62-3
IUPAC Name{2-[2-(dimethylamino)ethoxy]ethyl}dimethylamine
Molecular FormulaC8H20N2O
InChI KeyGTEXIOINCJRBIO-UHFFFAOYSA-N
SMILESCN(C)CCOCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.4280-1.4320 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only