N-Boc-L-prolinol, 98+%
N-Boc-L-prolinol, 98+%
N-Boc-L-prolinol, 98+%
Thermo Scientific Chemicals

N-Boc-L-prolinol, 98+%

CAS: 69610-40-8 | C10H19NO3 | 201.266 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L09885.03
also known as L09885-03
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.10
Save 7.45 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.10
Save 7.45 (14%)
Each
Chemical Identifiers
CAS69610-40-8
IUPAC Nametert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular FormulaC10H19NO3
InChI KeyBFFLLBPMZCIGRM-QMMMGPOBSA-N
SMILESCC(C)(C)OC(=O)N1CCC[C@H]1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 98.0%
Formcrystalline powder
Optical Rotation-54? ? 2? (c=5, methanol)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only