4-n-Butylphenyl isocyanate, 98%
4-n-Butylphenyl isocyanate, 98%
4-n-Butylphenyl isocyanate, 98%
Thermo Scientific Chemicals

4-n-Butylphenyl isocyanate, 98%

CAS: 69342-47-8 | C11H13NO | 175.231 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number L09957.03
also known as L09957-03
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.50
Save 4.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.50
Save 4.85
Each
Chemical Identifiers
CAS69342-47-8
IUPAC Name1-butyl-4-isocyanatobenzene
Molecular FormulaC11H13NO
InChI KeyLJJRXPXDTAUVQU-UHFFFAOYSA-N
SMILESCCCCC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.5145-1.5185 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only