Diethyl disulfide, 99%
Diethyl disulfide, 99%
Diethyl disulfide, 99%
Diethyl disulfide, 99%
Thermo Scientific Chemicals

Diethyl disulfide, 99%

CAS: 110-81-6 | C4H10S2 | 122.244 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L10065.14
also known as L10065-14
Price (USD)
53.65
Online Exclusive
59.50
Save 5.85 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.50
Save 5.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS110-81-6
IUPAC Name(ethyldisulfanyl)ethane
Molecular FormulaC4H10S2
InChI KeyCETBSQOFQKLHHZ-UHFFFAOYSA-N
SMILESCCSSCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)>98.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5045-1.5075 @ 20?C
Diethyl sulfide is used as a solvent for anhydrous minerals and in plating baths for gold and silver

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl sulfide is used as a solvent for anhydrous minerals and in plating baths for gold and silver

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Hiroshi Tsubomura.; Robert P. Lang. Molecular Complexes and Their Spectra. XIII. Complexes of Iodine with Amides, Diethyl Sulfide and Diethyl Disulfide.J. Am. Chem. Soc. 1961, 83 (9),2085-2092 .