Search
| Catalog Number | Quantity |
|---|---|
| L100CAL2 | 5 mL |
xMAP™ Reporter (CAL2) Calibrator microspheres are polystyrene microspheres that are internally labeled with fluorescent dyes. They serve as system Calibrators for Luminex xMAP technology based detectors (such as Luminex 200™) and are intended to normalize the reporter channel (RP1).
The Luminex™ 200™ System is sold separately.
For Research Use Only. Not for use in diagnostic procedures.