Product Image
Invitrogen™

xMAP™ Reporter Control Microspheres (CON2)

xMAP™ Reporter Control Microspheres (CON2), are polystyrene microspheres that are internally labeled with fluorescent dyes.
Have Questions?
Catalog NumberQuantity
L100CON25 mL
Catalog number L100CON2
Price (USD)
375.00
Each
Quantity:
5 mL
Price (USD)
375.00
Each

xMAP™ Reporter Control Microspheres (CON2), are polystyrene microspheres that are internally labeled with fluorescent dyes. These Control Microspheres are intended to verify the calibration and optical integrity for the Luminex 200™ System. Reporter Control Microspheres verify the reporter channel.

The Luminex™ 200™ System is sold separately.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Channel ClassificationReporter Channel (RP1)
For Use With (Equipment)Luminex™ Instruments, Luminex™ 200™ System
Product LineLuminex
Shipping ConditionWet Ice
Product TypeCalibration Microspheres
Quantity5 mL
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.