7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline, 97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline, 97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline, 97%
Thermo Scientific Chemicals

7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline, 97%

CAS: 450-62-4 | C10H10F3N | 201.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L10150.06
also known as L10150-06
Price (USD)
609.65
Online Exclusive
677.00
Save 67.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
609.65
Online Exclusive
677.00
Save 67.35 (10%)
Each
Chemical Identifiers
CAS450-62-4
IUPAC Name7-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline
Molecular FormulaC10H10F3N
InChI KeyRGZZKZNESVFQKR-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC2=C(CCCN2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or pale brown
FormCrystalline powder or fused solid
Melting Point (clear melt)29-34?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only